Anti-NWSHPQFEK Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Leverancier: Genscript
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

A00875-40
GENSA00875-40EA 173 EUR
GENSA00875-40
Anti-NWSHPQFEK Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antilichamen
Type: Tag
Antigen: NWSHPQFEK
Clonality: polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity:
Order Now

Specificaties

antigeen symbool NWSHPQFEK
Environmentally Preferable
Type antilichaam Label,NWSHPQFEK Tag
Type antilichaam Label,NWSHPQFEK Tag
Clonality Polyklonaal
Conjugatie HRP (Horseradish Peroxidase)
Reactivity
Host Rabbit
Isotype IgG
Western blot Yes
ELISA Yes
Opslagtemperatuur Cool
Shipping temperature Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR