Anti-NWSHPQFEK Tag Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase))

Leverancier: Genscript
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

A01742-100
GENSA01742-100EA 598 EUR
GENSA01742-100
Anti-NWSHPQFEK Tag Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase))
Antilichamen
Type: Tag
Antigen: NWSHPQFEK Tag
Clonality: monoclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR