Keyboard for medical environments, InduProof® Smart Touch
Leverancier: GETT GERATETECHNIK
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
KG22204 KG22205 KG23204 KG23213 KG23242 KG23254 KG23202 KG23220 KG23232 KG23000 KG25209
GETTKG22204EA 291 EUR
GETTKG22204 GETTKG22205 GETTKG23204 GETTKG23213 GETTKG23242 GETTKG23254 GETTKG23202 GETTKG23220 GETTKG23232 GETTKG23000 GETTKG25209
Keyboard for medical environments, InduProof® Smart Touch
Toetsenborden
Fully water-proof and dust-proof medical keyboard with NUM Block and touchpad.
  • Dust-proof and water-proof keyboard with integrated capacitive touchpad
  • Can be completely disinfected
  • Soft typing through silicone switching elements
  • Good basic quality and cost-effective
  • Flat key caps for thorough disinfection
  • Pleasant tactile feeling even with protective gloves
  • 3-year warranty on technical function
  • Made in Germany
This InduProof® keyboard has a complete set of keys with numeric keypad and an integrated touchpad. It offers a pleasant tactile feel and theMore Product Information