Waterproof and dustproof silicone mouse for hygienic areas, InduMouse® Pro
Leverancier: GETT GERATETECHNIK
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
KH25201 KH29200 KH29201
GETTKH25201EA 199 EUR
GETTKH25201 GETTKH29200 GETTKH29201
Waterproof and dustproof silicone mouse for hygienic areas, InduMouse® Pro
Computermuizen en muismatten
The InduMouse® Pro has an ergonomic design, a pleasant tactile feel, and an integrated scroll bar. With a high level of operating comfort and resistance to cleaning agents and chemicals, this mouse is an all-rounder in everyday medical use.
  • Versatile application possibilities
  • Symmetrical housing shape
  • IP68 protection rating
  • USB protection cap
  • Resolution: adjustable up to 1600 dpi
  • Customization options are available upon request
  • 4-year warranty on technical functionality (in addition to the statutory 2-year warranty)
Due to the waist, the InduMouse®More Product Information