Syringe filters, SupaTop
Leverancier: GILSON
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
ANM1322 ANM1322D ANM1345 ANM1345D ANM2522 ANM2522C ANM2545 ANM2545C ANN1322 ANN1322D ANN1345 ANN1345D ANN2522 ANN2522C ANN2545 ANN2545C ANP1322 ANP1322D ANP1345 ANP1345D ANP2522 ANP2522C ANP2545 ANP2545C ANR1322 ANR1322D ANR1345 ANR1345D ANR2522 ANR2522C ANR2545 ANR2545C ANV1322 ANV1322D ANV1345 ANV1345D ANV2522 ANV2522C ANV2545 ANV2545C
GILSANM1322EA 80 EUR
GILSANM1322 GILSANM1322D GILSANM1345 GILSANM1345D GILSANM2522 GILSANM2522C GILSANM2545 GILSANM2545C GILSANN1322 GILSANN1322D GILSANN1345 GILSANN1345D GILSANN2522 GILSANN2522C GILSANN2545 GILSANN2545C GILSANP1322 GILSANP1322D GILSANP1345 GILSANP1345D GILSANP2522 GILSANP2522C GILSANP2545 GILSANP2545C GILSANR1322 GILSANR1322D GILSANR1345 GILSANR1345D GILSANR2522 GILSANR2522C GILSANR2545 GILSANR2545C GILSANV1322 GILSANV1322D GILSANV1345 GILSANV1345D GILSANV2522 GILSANV2522C GILSANV2545 GILSANV2545C
Syringe filters, SupaTop
Filters Spuitfilters
SupaTop syringe filters offer reliable and cost-effective solutions for particles removal and clarification needs.
  • Body made of pigment-free PP
  • Certified for low levels of UV-absorbing extractables
A large variety of membranes are available for all applications: MCE, Nylon, PTFE, PES or PVDF.