Pipette stand for Gilson pipettes, TRIO™
Leverancier: GILSON
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
F161405
GILSF161405EA 111 EUR
GILSF161405
Pipette stand for Gilson pipettes, TRIO™
Pipethouders
This handy stand with its angled, transparent design holds up to three single or multi channel Gilson pipettes.
  • Made of Temperlite material: Very shock resistant, can be used in chemically aggressive environments
  • Protects valuable instruments from shocks and contamination
  • Pipettes hang vertically to prevent liquids from running inside the shaft
  • Maintain visibility: Its transparent body allows users to see the objects placed behind
  • Space saving for a neat, tidy and organized work bench