Filtered and sterilised tips, PIPETMAN® EXPERT
Leverancier: GILSON
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
F1732031 F1733031 F1734031 F1735031 F1735041 F1736031 F1737031 F1731032 F1732032 F1733032 F1734032 F1735032 F1735042 F1736032 F1737032 F1731031
GILSF1732031EA 125 EUR
GILSF1732031 GILSF1733031 GILSF1734031 GILSF1735031 GILSF1735041 GILSF1736031 GILSF1737031 GILSF1731032 GILSF1732032 GILSF1733032 GILSF1734032 GILSF1735032 GILSF1735042 GILSF1736032 GILSF1737032 GILSF1731031
Filtered and sterilised tips, PIPETMAN® EXPERT
Pipettips
PIPETMAN® EXPERT Tips are designed to fit leading pipette brands, allowing you to stock one brand of tips saving time, money, and bench space. EXPERT Filter Tips feature a patented, low-retention polymer that creates a hydrophobic surface, ensuring complete dispensing of the sample for more accurate results. The tips are equipped with a polyethylene filter to prevent aerosol-born contamination.
  • Low-retention tips for complete dispensing
  • Free of detectable contaminants
  • Convenient, universal fit tips
The low-retention polymer coating prevents samplesMore Product Information