Petrischalen, Corning® Gosselin™
Leverancier: Corning
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
BP140-01 BP140-02 BP53-03 BP53-06 BP93D-102 SB93-101 BH93B-102 BI93D-101 BI93D-102 BP90B-101 BP90B-102 BP93B-101 SB93-102 BP143-06 BP143-07 BP93B-102
391-2068EA 224 EUR
391-2068 GOSSBP140-02 391-0569 391-0570 GOSSBP93D-102 391-0572 391-0566 391-0567 391-0568 391-0571 GOSSBP90B-102 GOSSBP93B-101 GOSSSB93-102 GOSSBP143-06 GOSSBP143-07 GOSSBP93B-102
Petrischalen, Corning® Gosselin™
Cultuurschalen Petrischalen
Transparant PS. Petri schalen voor levensmiddelenindustrie of laboratoria.
  • Uitstekende optische kwaliteit
  • Stabiel stapelbaar door de stapelring
  • Hoge mechanische bestendigheid tot +55 °C
These dishes are produced in a Class 100 (ISO 5) manufacturing environment, ensuring an aseptic product. Product is labelled with a batch number on each carton. Also available sterile (Sterility Assurance Level based on ISO 11137 : SAL 10 ⁻⁶).