pH/mV/°C/rH meter, handheld, GMH 3531
Leverancier: Greisinger
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
602076
GRSG602076EA 0 EUR
GRSG602076
pH/mV/°C/rH meter, handheld, GMH 3531
Elektrochemische meters pH- en multiparametersystemen
This meter is a simple handheld instrument for measuring pH, mV, temperature and relative humidity (0,0 to 70,0).
  • Battery operation makes it ideal for field measurements
  • Min./Max. value and HOLD memory
  • Impact-resistant ABS plastic housing, membrane keyboard, transparent panel; front side with IP 65 rating
  • Integrated pop-up clip for table top or suspended use
  • Serial interface (3,5 mm jack) can be connected to RS232 or USB interface of a PC via electrically isolated interface converter GRS 3100, GRS 3105 or USB 3100 N (available separately)
  • ORP mode allows
More Product Information