2-Bromo-6-(trifluoromethyl)pyridine ≥97%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

H25749.03 H25749.06
H25749.03EA 114 EUR
H25749.03 H25749.06
2-Bromo-6-(trifluoromethyl)pyridine ≥97%
2-Bromo-6-(trifluoromethyl)pyridine

Purity: 97% CAS No.: 189278-27-1 Molecular Formula: C6H3BrF3N Formula Weight: 226.00 Melting Point: 48-51° Boiling Point: 78-79°/14mm MDL No.: MFCD00153087

Formule: C₆H₃BrF₃N
Kookpunt: 78…79 °C (14 mmHg)
Smeltpunt: 48…51 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00153087
CAS nummer: 189278-27-1

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR