2-Fluoroisonicotinonitrile ≥97%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

H25938.14 H25938.06 H25938.03
H25938.14EA 166 EUR
H25938.14 H25938.06 H25938.03
2-Fluoroisonicotinonitrile ≥97%
2-Fluoroisonicotinonitrile

Purity: 97% CAS No.: 3939-14-8 Molecular Formula: C6H3FN2 Formula Weight: 122.10 Melting Point: 31-33° MDL No.: MFCD09027299

Formule: C₆H₃FN₂
Smeltpunt: 31…33 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD09027299
CAS nummer: 3939-14-8

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR