5,10,15,20-Tetrakis(4-methoxyphenyl)porphyrin 95%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

H25946.09 H25946.03
H25946.09EA 306 EUR
H25946.09 H25946.03
5,10,15,20-Tetrakis(4-methoxyphenyl)porphyrin 95%
5,10,15,20-Tetrakis(4-methoxyphenyl)porphyrin

Purity: 95% CAS No.: 22112-78-3 Molecular Formula: C48H38N4O4 Formula Weight: 734.87 Storage Temperature: 2°C to 8°C MDL No.: MFCD09909566

Formule: C₄₈H₃₈N₄O₄
MDL Nummer: MFCD00012072
CAS nummer: 22112-78-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR