4-Amino-2-fluorobenzoic acid 98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

H26146.03
H26146.03EA 132 EUR
H26146.03
4-Amino-2-fluorobenzoic acid 98%
4-Amino-2-fluorobenzoic acid

Purity: 98% CAS No.: 446-31-1 Molecular Formula: C7H6FNO2 Formula Weight: 155.13 Melting Point: 209-211° MDL No.: MFCD01569397

Formule: C₇H₆FNO₂
Smeltpunt: 209…211 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD01569397
CAS nummer: 446-31-1
EINECS: None

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR