Dimethyl (4-fluorobenzyl)malonate 97%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

H26310.03 H26310.06
H26310.03EA 41 EUR
H26310.03 H26310.06
Dimethyl (4-fluorobenzyl)malonate 97%
Dimethyl (4-fluorobenzyl)malonate

Purity: 97% CAS No.: 252938-22-0 Molecular Formula: C12H13FO4 Formula Weight: 240.23 MDL No.: MFCD01934558

Formule: C₁₂H₁₃FO₄
MW: 240.23 g/mol
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD01934558
CAS nummer: 252938-22-0
EINECS: None

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR