N-Boc-L-prolinal 96%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

H26495.06 H26495.03
H26495.06EA 428 EUR
H26495.06 H26495.03
N-Boc-L-prolinal 96%
N-Boc-L-prolinal

Purity: 96% CAS No.: 69610-41-9 Molecular Formula: C10H17NO3 Formula Weight: 199.25 Storage Temperature: -30°C to -10°C Flash Point: 110°(230°F) Refractive Index: 1.4589 Boiling Point: 228° Optical Rotation: -95° (c=1 in chloroform) MDL No.: MFCD00274186

Formule: C₁₀H₁₇NO₃
Kookpunt: 228 °C
Dichtheid: 1,06
Vlampunt: >110 °C (230 °F)
MDL Nummer: MFCD00274186
CAS nummer: 69610-41-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR