3-Iodopyridine-2-carbonitrile 95%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

H26963.MD H26963.03
H26963.MDEA 82.7 EUR
H26963.MD H26963.03
3-Iodopyridine-2-carbonitrile 95%
3-Iodopyridine-2-carbonitrile

Purity: 95% CAS No.: 827616-52-4 Molecular Formula: C6H3IN2 Formula Weight: 230.01 Melting Point: 98-100° UN No.: 3439 MDL No.: MFCD09037451

Formule: C₆H₃IN₂
Smeltpunt: 98…100 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD09037451
CAS nummer: 827616-52-4
UN: 3439
ADR: 6.1,III

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR