1-Butyl-3-methylpyridiniumtetrafluoroborate ≥99%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

H26983.18 H26983.06
H26983.18EA 222 EUR
H26983.18 H26983.06
1-Butyl-3-methylpyridiniumtetrafluoroborate ≥99%
1-Butyl-3-methylpyridiniumtetrafluoroborate

Purity: 99% CAS No.: 597581-48-1 Molecular Formula: C10H16BF4N Formula Weight: 237.06 MDL No.: MFCD08458499

Formule: C₁₀H₁₆BF₄N
Dichtheid: 1,204
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD08458499
CAS nummer: 597581-48-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR