Ammoniak 2 M in methanol

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

H27080.AUEA 280 EUR
H27080.AU H27080.AE
Ammoniak 2 M in methanol
Ammoniak

CAS No.: 7664-41-7 Molecular Formula: H3N Formula Weight: 17.03 Storage Temperature: 2°C to 8°C Flash Point: 6°(43°F) MDL No.: MFCD00011418

Formule: NH₃
MW: 17.03 g/mol
Kookpunt: -33
Smeltpunt: -77
Dichtheid: 0.787
Vlampunt: 6 °C (43 °F)
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00011418
CAS nummer: 7664-41-7
EINECS: 215-647-6
UN: 2924
ADR: 3 II
Merck Index: 14,492

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR