5-Pyrimidineboronic acid pinacol ester 95%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

H27147.03
H27147.03EA 102 EUR
H27147.03
5-Pyrimidineboronic acid pinacol ester 95%
5-Pyrimidineboronic acid pinacol ester

Purity: 95% CAS No.: 321724-19-0 Molecular Formula: C10H15BN2O2 Formula Weight: 206.05 Storage Temperature: 2°C to 8°C Melting Point: 60-66° MDL No.: MFCD05155223

Formule: C₁₀H₁₅BN₂O₂
Smeltpunt: 60…66 °C
MDL Nummer: MFCD05155223
CAS nummer: 321724-19-0

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR