n-Pentaan 99+% HPLC

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: Pentane

H27427.AU H27427.0F
H27427.AUEA 73.7 EUR
H27427.AU H27427.0F
n-Pentaan 99+% HPLC
n-Pentaan

Purity: 99+% CAS No.: 109-66-0 Molecular Formula: CH3(CH2)3CH3 Formula Weight: 72.15 Flash Point: -49°(-57°F) Refractive Index: 1.3580 Odour: Nearly Odourless Melting Point: -130° Boiling Point: 36° Merck Reference: 14,7116 UN No.: 1265 MDL No.: MFCD00009498

Formule: H₃C(CH₂)₃CH₃
MW: 72.15 g/mol
Kookpunt: 36 °C
Smeltpunt: –130 °C
Dichtheid: 0.626
Vlampunt: –49 °C (–57 °F)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00009498
CAS nummer: 109-66-0
EINECS: 203-692-4
UN: 1265
ADR: 3 II
Bereik: 01-2119459286-30
Merck Index: 14,7116
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4ba2c9c81a4b1f44769627a00ba49b6cc8e63132&vid=f6831eb15ab1602d8c752468dc3ee31cc4ee61c0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance (Color) Clear, colorless
Form Liquid
Assay (unspecified) (all isomers) >99.5%
Assay (unspecified) (n-Pentane) >95%
Water Content (Karl Fischer Titration) < 50 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR