2-Amino-3-nitrophenol 98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

H27472.14 H27472.06
H27472.14EA 234 EUR
H27472.14 H27472.06
2-Amino-3-nitrophenol 98%
2-Amino-3-nitrophenol

Purity: 98% CAS No.: 603-85-0 Molecular Formula: C6H6N2O3 Formula Weight: 154.13 Melting Point: ca 212° Beilstein Reference: 2804307

Formule: C₆H₆N₂O₃
Smeltpunt: ∼ 212 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00010875
CAS nummer: 603-85-0
EINECS: 210-060-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR