Cyclopropylboronic acid pinacol ester 96%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

H27642.06 H27642.03
H27642.06EA 139 EUR
H27642.06 H27642.03
Cyclopropylboronic acid pinacol ester 96%
Cyclopropylboronic acid pinacol ester

Purity: 96% CAS No.: 126689-01-8 Molecular Formula: C9H17BO2 Formula Weight: 168.04 Storage Temperature: -30°C to -10°C Flash Point: 40°(104°F) Boiling Point: 146° UN No.: 3272 MDL No.: MFCD05663847

Formule: C₉H₁₇BO₂
Kookpunt: 146 °C
Dichtheid: 0,922
Vlampunt: 40 °C (104 °F)
MDL Nummer: MFCD05663847
CAS nummer: 126689-01-8
UN: 3272
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR