Dimethylamine 2 M THF

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

H27665.AEEA 85.1 EUR
H27665.AE H27665.AU
Dimethylamine 2 M THF
Dimethylamine

CAS No.: 124-40-3 Molecular Formula: C2H7N Formula Weight: 45.08 Flash Point: -36°(-33°F) Boiling Point: ca 60° Beilstein Reference: 605257 Merck Reference: 14,3228 UN No.: 2924 MDL No.: MFCD00008288

Formule: C₂H₇N
MW: 45.08 g/mol
Kookpunt: ∼60 °C
Smeltpunt: -92.8
Dichtheid: 0.85
Vlampunt: –36 °C (–33 °F)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00008288
CAS nummer: 124-40-3
EINECS: 204-697-4
UN: 2924
ADR: 3 II
Merck Index: 14,3228

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR