N-Boc-D-phenylalaninol ≥98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

H27713.03 H27713.06
H27713.03EA 54.6 EUR
H27713.03 H27713.06
N-Boc-D-phenylalaninol ≥98%
N-Boc-D-phenylalaninol

Purity: 98% CAS No.: 106454-69-7 Molecular Formula: C14H21NO3 Formula Weight: 251.33 Melting Point: 94-96° MDL No.: MFCD00216472

Formule: C₁₄H₂₁NO₃
Smeltpunt: 94…96 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00216472
CAS nummer: 106454-69-7

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR