Methyl oleate 96%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: cis-Oleic Acid Methyl Ester, Oleic acid methyl ester

H31358.14 H31358.22
H31358.14EA 144 EUR
H31358.14 H31358.22
Methyl oleate 96%
Methyl oleate

Purity: 96% CAS No.: 112-62-9 Molecular Formula: C19H36O2 Formula Weight: 296.50 Flash Point: 163°(325°F) Refractive Index: 1.4520 Boiling Point: 168-170°/2mm Beilstein Reference: 1727037 Merck Reference: 14,6828 MDL No.: MFCD00009578

Formule: C₁₉H₃₆O₂
MW: 296.49 g/mol
Kookpunt: 168…170 °C (2 mmHg)
Smeltpunt: –20 °C
Dichtheid: 0.876
Vlampunt: 163 °C (325 °F)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00009578
CAS nummer: 112-62-9
EINECS: 203-992-5
Merck Index: 12,06965

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR