8-Hydroxyjulolidine-9-carboxaldehyde 97%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

H31782.03 H31782.06
H31782.03EA 57.4 EUR
H31782.03 H31782.06
8-Hydroxyjulolidine-9-carboxaldehyde 97%
8-Hydroxyjulolidine-9-carboxaldehyde

Purity: 97% CAS No.: 63149-33-7 Molecular Formula: C13H15NO2 Formula Weight: 217.27 Melting Point: 72-74° MDL No.: MFCD00192477

Formule: C₁₃H₁₅NO₂
Smeltpunt: 72…74 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00192477
CAS nummer: 63149-33-7
EINECS: 263-966-4

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR