Thionylchloride 1 M in dichloromethane

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: Thionyl dichloride

H31857.AE H31857.AP
H31857.AEEA 54.4 EUR
H31857.AE H31857.AP
Thionylchloride 1 M in dichloromethane
Thionylchloride

CAS No.: 7719-09-7 Molecular Formula: Cl2OS Formula Weight: 118.97 UN No.: 2922 MDL No.: MFCD00011449

Formule: Cl₂OS
Dichtheid: 1,35
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011449
CAS nummer: 7719-09-7
EINECS: 231-748-8
UN: 2922
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR