Ethyl-2-amino-4-phenylthiophene-3-carboxylate 96%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

H31867.03 H31867.09
H31867.03EA 23.6 EUR
H31867.03 H31867.09
Ethyl-2-amino-4-phenylthiophene-3-carboxylate 96%
Ethyl-2-amino-4-phenylthiophene-3-carboxylate

Purity: 96% CAS No.: 4815-36-5 Molecular Formula: C13H13NO2S Formula Weight: 247.32 Physical Form: Crystalline solid Appearance: Yellow Melting Point: 90-94° MDL No.: MFCD00126391

Formule: C₁₃H₁₃NO₂S
Smeltpunt: 90…94 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00126391
CAS nummer: 4815-36-5

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR