2,6-Difluoro-3-hydroxybenzaldehyde 95%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

H32250.MD H32250.03
H32250.MDEA 74.4 EUR
H32250.MD H32250.03
2,6-Difluoro-3-hydroxybenzaldehyde 95%
2,6-Difluoro-3-hydroxybenzaldehyde

Purity: 95% CAS No.: 152434-88-3 Molecular Formula: C7H4F2O2 Formula Weight: 158.11 Melting Point: 110-114° MDL No.: MFCD08275284

Formule: C₇H₄F₂O₂
Smeltpunt: 110…114 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD08275284
CAS nummer: 152434-88-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR