4-Bromo-3-fluorophenol ≥98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

H32488.06 H32488.14
H32488.06EA 54.2 EUR
H32488.06 H32488.14
4-Bromo-3-fluorophenol ≥98%
4-Bromo-3-fluorophenol

Purity: 98% CAS No.: 121219-03-2 Molecular Formula: C6H4BrFO Formula Weight: 191.00 Melting Point: 71-73° MDL No.: MFCD00051907

Formule: C₆H₄BrFO
Smeltpunt: 71…73 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00051907
CAS nummer: 121219-03-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR