Trimethylamine 1 M in THF

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

H32899.AE H32899.AP
H32899.AEEA 94.6 EUR
H32899.AE H32899.AP
Trimethylamine 1 M in THF
Trimethylamine

CAS No.: 75-50-3 Molecular Formula: C3H9N Formula Weight: 59.11 UN No.: 1993 MDL No.: MFCD00008327

Formule: C₃H₉N
Dichtheid: 0,850
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00008327
CAS nummer: 75-50-3
EINECS: 200-875-0
UN: 2924
ADR: 3,II

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR