2-Bromo-4-(trifluoromethyl)benzoic acid ≥98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

H32989.03 H32989.06
H32989.03EA 103 EUR
H32989.03 H32989.06
2-Bromo-4-(trifluoromethyl)benzoic acid ≥98%
2-Bromo-4-(trifluoromethyl)benzoic acid

Purity: 98% CAS No.: 328-89-2 Molecular Formula: C8H4BrF3O2 Formula Weight: 269.02 MDL No.: MFCD07368837

Formule: C₈H₄BrF₃O₂
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD07368837
CAS nummer: 328-89-2
EINECS: 206-339-2

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR