Isopropylboronic acid pinacol ester ≥98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

H33861.03 H33861.06 H33861.14
H33861.03EA 23.4 EUR
H33861.03 H33861.06 H33861.14
Isopropylboronic acid pinacol ester ≥98%
Isopropylboronic acid pinacol ester
Formule: C₉H₁₉BO₂
Kookpunt: 115 °C (60 mmHg)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD05663856
CAS nummer: 76347-13-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR