4'-Methylbiphenyl-4-carboxylic acid 96%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

H34459.03 H34459.06
H34459.03EA 59.2 EUR
H34459.03 H34459.06
4'-Methylbiphenyl-4-carboxylic acid 96%
4'-Methylbiphenyl-4-carboxylic acid
Formule: C₁₄H₁₂O₂
Smeltpunt: 257…258 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00448778
CAS nummer: 720-73-0

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR