(R)-(+)-2-Hydroxy-3-phenylpropionic acid ≥98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

H35096.03 H35096.06
H35096.03EA 24.5 EUR
H35096.03 H35096.06
(R)-(+)-2-Hydroxy-3-phenylpropionic acid ≥98%
(R)-(+)-2-Hydroxy-3-phenylpropionic acid
Formule: C₉H₁₀O₃
MDL Nummer: MFCD00078062
CAS nummer: 7326-19-4
EINECS: 230-803-3

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR