Benzo[b]thiophene-3-carboxylic acid 96%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

H50105.03 H50105.06 H50105.14
H50105.03EA 70.5 EUR
H50105.03 H50105.06 H50105.14
Benzo[b]thiophene-3-carboxylic acid 96%
Benzo[b]thiophene-3-carboxylic acid

Purity: 96% CAS No.: 5381-25-9 Molecular Formula: C9H6O2S Formula Weight: 178.22 Melting Point: 174-176° MDL No.: MFCD01846406

Formule: C₉H₆O₂S
Smeltpunt: 174…176 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD01846406
CAS nummer: 5381-25-9

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR