2-(Cyclopropylmethoxy)-5-pyridineboronic acid pinacol ester ≥97%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

H50139.03 H50139.MD
H50139.03EA 121 EUR
H50139.03 H50139.MD
2-(Cyclopropylmethoxy)-5-pyridineboronic acid pinacol ester ≥97%
2-(Cyclopropylmethoxy)-5-pyridineboronic acid pinacol ester

Purity: 97% CAS No.: 947191-69-7 Molecular Formula: C15H22BNO3 Formula Weight: 275.16 MDL No.: MFCD07781184

Formule: C₁₅H₂₂BNO₃
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD07781184
CAS nummer: 947191-69-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR