Isopropyl glycolate 96%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

H50334.03 H50334.06
H50334.03EA 91.3 EUR
H50334.03 H50334.06
Isopropyl glycolate 96%
Isopropyl glycolate

Purity: 96% CAS No.: 623-61-0 Molecular Formula: HOCH2CO2CH(CH3)2 Formula Weight: 118.13 MDL No.: MFCD00046747

Formule: HOCH₂CO₂CH(CH₃)₂
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00046747
CAS nummer: 623-61-0
EINECS: 210-804-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR