1-Methyl-2-(tributylstannyl)pyrrole ≥98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

H51438.03 H51438.06
H51438.03EA 96 EUR
H51438.03 H51438.06
1-Methyl-2-(tributylstannyl)pyrrole ≥98%
1-Methyl-2-(tributylstannyl)pyrrole

Purity: 98% CAS No.: 118486-97-8 Molecular Formula: C17H33NSn Formula Weight: 370.15 Flash Point: 98°(208°F) Refractive Index: 1.511 UN No.: 2788 MDL No.: MFCD01319022

Formule: C₁₇H₃₃NSn
Dichtheid: 1,122
Vlampunt: 98 °C (208 °F)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD01319022
CAS nummer: 118486-97-8
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR