Diethyl (3-aminopropyl)phosphonate 95%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

H51874.MD H51874.03
H51874.MDEA 136 EUR
H51874.MD H51874.03
Diethyl (3-aminopropyl)phosphonate 95%
Diethyl (3-aminopropyl)phosphonate

Purity: 95% CAS No.: 4402-24-8 Molecular Formula: C7H18NO3P Formula Weight: 195.20 Storage Temperature: 2°C to 8°C

Formule: C₇H₁₈NO₃P
MW: 195.20 g/mol
Dichtheid: 1.053
MDL Nummer: MFCD11110596
CAS nummer: 4402-24-8
EINECS: 224-535-6
ADR: 8 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR