Fmoc-L-β-homoaspartic acid-gamma-t-butyl ester 95%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

H52190.MD H52190.03
H52190.MDEA 155 EUR
H52190.MD H52190.03
Fmoc-L-β-homoaspartic acid-gamma-t-butyl ester 95%
Fmoc-L-beta-homoaspartic acid-gamma-t-butyl ester

Purity: 95% CAS No.: 209252-17-5 Molecular Formula: C24H27NO6 Formula Weight: 425.48 Beilstein Reference: 7952342 MDL No.: MFCD01862860

Formule: C₂₄H₂₇NO₆
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD01862860
CAS nummer: 209252-17-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR