3-Azetidinecarboxylic acid ≥98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

H52294.03 H52294.06
H52294.03EA 120 EUR
H52294.03 H52294.06
3-Azetidinecarboxylic acid ≥98%
3-Azetidinecarboxylic acid

Purity: 98+% CAS No.: 36476-78-5 Molecular Formula: C4H7NO2 Formula Weight: 101.11 Melting Point: ca 250° dec. MDL No.: MFCD00191763

Formule: C₄H₇NO₂
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00191763
CAS nummer: 36476-78-5

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR