4-Methoxyindole-2-carboxylic acid ≥97%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

H52392.MD H52392.03 H52392.06
H52392.MDEA 47.4 EUR
H52392.MD H52392.03 H52392.06
4-Methoxyindole-2-carboxylic acid ≥97%
4-Methoxyindole-2-carboxylic acid

Purity: 97+% CAS No.: 103260-65-7 Molecular Formula: C10H9NO3 Formula Weight: 191.19 MDL No.: MFCD02664458

Formule: C₁₀H₉NO₃
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD02664458
CAS nummer: 103260-65-7

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR