(R)-3-(Boc-amino)piperidine 97%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

H52749.03 H52749.06
H52749.03EA 50.2 EUR
H52749.03 H52749.06
(R)-3-(Boc-amino)piperidine 97%
(R)-3-(Boc-amino)piperidine

Purity: 97% CAS No.: 309956-78-3 Molecular Formula: C10H20N2O2 Formula Weight: 200.28 MDL No.: MFCD03093382

Formule: C₁₀H₂₀N₂O₂
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD03093382
CAS nummer: 309956-78-3

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR