tert-Butyl-4-oxoazepane-1-carboxylate 98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

H55214.03 H55214.06
H55214.03EA 92.5 EUR
H55214.03 H55214.06
tert-Butyl-4-oxoazepane-1-carboxylate 98%
tert-Butyl-4-oxoazepane-1-carboxylate

Purity: 98% CAS No.: 188975-88-4 Molecular Formula: C11H19NO3 Formula Weight: 213.28 Storage Temperature: 2°C to 8°C Refractive Index: 1.4670 MDL No.: MFCD03788435

Formule: C₁₁H₁₉NO₃
MDL Nummer: MFCD03788435
CAS nummer: 188975-88-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR