N-Boc-4-bromoaniline ≥97%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

H55226.06
H55226.06EA 86.7 EUR
H55226.06
N-Boc-4-bromoaniline ≥97%
N-Boc-4-bromoaniline
Formule: C₁₁H₁₄BrNO₂
Smeltpunt: 103…106 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD01006612
CAS nummer: 131818-17-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR