9,9-Di-n-dodecyl-2,7-dibromofluorene 98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

H56320.14 H56320.22
H56320.14EA 157 EUR
H56320.14 H56320.22
9,9-Di-n-dodecyl-2,7-dibromofluorene 98%
9,9-Di-n-dodecyl-2,7-dibromofluorene

Purity: 98% CAS No.: 286438-45-7 Molecular Formula: C37H56Br2 Formula Weight: 660.65 Flash Point: 110°(230°F) Melting Point: 50-55° Boiling Point: 180°/0.15mm MDL No.: MFCD03427215

Formule: C₃₇H₅₆Br₂
Kookpunt: 180 °C (0,15 mmHg)
Smeltpunt: 50…55 °C
Vlampunt: 110 °C (230 °F)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD03427215
CAS nummer: 286438-45-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR