DSG (Disuccinimidyl glutarate) 97%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: DSG

H58208.MCEA 166 EUR
H58208.MC H58208.ME
DSG (Disuccinimidyl glutarate) 97%
DSG (Disuccinimidyl glutarate)

Purity: 97% CAS No.: 79642-50-5 Molecular Formula: C13H14N2O8 Formula Weight: 326.26 Storage Temperature: -30°C to -10°C Beilstein Reference: 8575145 MDL No.: MFCD00153597

Formule: C₁₃H₁₄N₂O₈
MW: 326.26 g/mol
MDL Nummer: MFCD00153597
CAS nummer: 79642-50-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR