FMOC-O-tert-butyl-L-tyrosine 98% (by HPLC)

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

H59730.06 H59730.14
H59730.06EA 60.5 EUR
H59730.06 H59730.14
FMOC-O-tert-butyl-L-tyrosine 98% (by HPLC)
FMOC-O-tert-butyl-L-tyrosine
Formule: C₂₈H₂₉NO₅
Smeltpunt: 153…156 °C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00037129
CAS nummer: 71989-38-3
EINECS: 276-262-7

Order Now

Specification Test Results

Assay (HPLC) >97.5%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR