Potassium cyclopropyltrifluoroborate ≥90%, tech.

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

H60673.03 H60673.06
H60673.03EA 81.4 EUR
H60673.03 H60673.06
Potassium cyclopropyltrifluoroborate ≥90%, tech.
Potassium cyclopropyltrifluoroborate
Formule: C₃H₅BF₃K
Smeltpunt: 348…350 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD09265031
CAS nummer: 1065010-87-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR